Assay Details
Target Gene Details
Entrez Gene ID: | 28960 |
Gene Name: | decapping enzyme, scavenger |
Gene Aliases: |
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015 |
Location: |
Chr.11:126303301-126345754 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCPS | NM_014026.4 | NP_054745.1 | ||
| XM_011542778.2 | XP_011541080.1 | |||
| XM_017017612.1 | XP_016873101.1 | |||
| AF077201.1 | AAD26996.1 | |||
| AF532613.1 | AAM90310.1 | |||
| AK026554.1 | ||||
| AK223089.1 | BAD96809.1 | |||
| AK307403.1 | ||||
| AY040771.1 | AAK91765.1 | |||
| AY077684.1 | AAL77216.1 | |||
| BC014532.1 | AAH14532.1 | |||
| BE382749.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556530 | Chr.11:126264724 - 126393990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map