Assay Details
Target Gene Details
Entrez Gene ID: | 79739 |
Gene Name: | tubulin tyrosine ligase like 7 |
Gene Aliases: |
- |
Location: |
Chr.1:83865021-83999340 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL7 | NM_024686.4 | NP_078962.4 | ||
| XM_005271208.2 | 20 | 3482 | XP_005271265.1 | |
| XM_017002347.1 | 22 | 3691 | XP_016857836.1 | |
| XM_017002349.1 | 20 | 3446 | XP_016857838.1 |
More Information
Set Membership: |
|

Genomic Map