Assay Details
Target Gene Details
Entrez Gene ID: | 10491 |
Gene Name: | cartilage associated protein |
Gene Aliases: |
CASP, LEPREL3, OI7, P3H5 |
Location: |
Chr.3:33113958-33147773 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTAP | NM_006371.4 | NP_006362.1 | ||
| AJ006470.1 | CAA07054.1 | |||
| AK001634.1 | ||||
| AK075401.1 | ||||
| AK091772.1 | BAC03743.1 | |||
| AK092990.1 | ||||
| AK097650.1 | ||||
| AK126433.1 | ||||
| AK298250.1 | ||||
| AK314719.1 | ||||
| BC008745.2 | AAH08745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map