Assay Details
Target Gene Details
Entrez Gene ID: | 1946 |
Gene Name: | ephrin A5 |
Gene Aliases: |
AF1, EFL5, EPLG7, GLC1M, LERK7, RAGS |
Location: |
Chr.5:107376889-107672364 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNA5 | NM_001962.2 | 5 | 1318 | NP_001953.1 |
| XM_006714565.2 | 4 | 2706 | XP_006714628.1 | |
| XM_011543250.2 | 5 | 1131 | XP_011541552.1 | |
| XM_011543251.2 | 5 | 2644 | XP_011541553.1 | |
| XM_017009205.1 | 5 | 2407 | XP_016864694.1 | |
| U26403.1 | 5 | 1318 | AAB60377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017067 | Chr.5:106985082 - 108550974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map