Assay Details
Target Gene Details
Entrez Gene ID: | 79595 |
Gene Name: | Sin3A associated protein 130 |
Gene Aliases: |
- |
Location: |
Chr.2:127941217-128028093 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAP130 | NM_001145928.1 | 21 | 3674 | NP_001139400.1 |
| NM_024545.3 | 20 | 3569 | NP_078821.2 | |
| XM_005263767.3 | 21 | 3645 | XP_005263824.1 | |
| XM_005263768.3 | 21 | 3624 | XP_005263825.1 | |
| XM_005263769.3 | 21 | 3545 | XP_005263826.1 | |
| XM_005263770.3 | 21 | 3621 | XP_005263827.1 | |
| XM_006712748.3 | 21 | 3831 | XP_006712811.1 | |
| XM_006712749.3 | 20 | 3726 | XP_006712812.1 | |
| XM_017004903.1 | 21 | 3621 | XP_016860392.1 | |
| XM_017004904.1 | 20 | 3519 | XP_016860393.1 | |
| XM_017004905.1 | 20 | 3516 | XP_016860394.1 | |
| AK022823.1 | 11 | 2191 | BAB14261.1 | |
| AK092234.1 | 14 | 2627 | ||
| AL136833.1 | 20 | 3567 | CAB66767.1 | |
| BC017453.1 | 13 | 2374 | AAH17453.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508865 | Chr.2:127888968 - 127971794 on Build GRCh38 | Insertion |
|
| nsv834367 | Chr.2:127873217 - 128032498 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map