Assay Details
Target Gene Details
Entrez Gene ID: | 10516 |
Gene Name: | fibulin 5 |
Gene Aliases: |
ADCL2, ARCL1A, ARMD3, DANCE, EVEC, FIBL-5, HNARMD, UP50 |
Location: |
Chr.14:91869411-91947702 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBLN5 | NM_006329.3 | 11 | 2512 | NP_006320.2 |
| XM_005267267.3 | 11 | 2347 | XP_005267324.1 | |
| XM_011536356.1 | 12 | 2526 | XP_011534658.1 | |
| XM_011536357.1 | 12 | 2681 | XP_011534659.1 | |
| XM_011536358.1 | 13 | 2781 | XP_011534660.1 | |
| XM_017020929.1 | 12 | 2602 | XP_016876418.1 | |
| AA812956.1 | 1 | 126 | ||
| AF112152.1 | 11 | 2207 | AAD41768.1 | |
| AJ133490.1 | CAB38568.1 | |||
| AK074540.1 | 5 | 1418 | ||
| AK094032.1 | 1 | 2202 | ||
| AK125600.1 | 2 | 2174 | ||
| AL110148.1 | 1 | 511 | ||
| AY358898.1 | 11 | 2488 | AAQ89257.1 | |
| BC022280.1 | 11 | 2222 | AAH22280.1 | |
| BX248290.1 | 12 | 2332 | CAD62618.1 | |
| BX537531.1 | 1 | 6536 |
More Information
Set Membership: |
|

Genomic Map