Assay Details
Target Gene Details
Entrez Gene ID: | 55716 |
Gene Name: | limb development membrane protein 1 like |
Gene Aliases: |
LIMR |
Location: |
Chr.12:49096551-49111535 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBR1L | NM_001300750.1 | 18 | 2459 | NP_001287679.1 |
| NM_001300751.1 | 17 | 2197 | NP_001287680.1 | |
| NM_018113.3 | 17 | 2257 | NP_060583.2 | |
| XM_011538563.2 | 17 | 2411 | XP_011536865.1 | |
| XM_011538564.2 | 17 | 2784 | XP_011536866.1 | |
| XM_011538565.2 | 16 | 3033 | XP_011536867.1 | |
| XM_011538566.2 | 16 | 2007 | XP_011536868.1 | |
| XM_017019626.1 | 17 | 2659 | XP_016875115.1 | |
| XM_017019627.1 | 18 | 2388 | XP_016875116.1 | |
| XM_017019628.1 | 16 | 2084 | XP_016875117.1 | |
| XM_017019629.1 | 18 | 3243 | XP_016875118.1 | |
| XM_017019630.1 | 14 | 4219 | XP_016875119.1 | |
| XM_017019631.1 | 16 | 1947 | XP_016875120.1 | |
| AB033000.1 | 15 | 2728 | BAA86488.1 | |
| AF260728.1 | 17 | 2216 | AAK60600.1 | |
| AK001356.1 | 18 | 2240 | BAA91646.1 | |
| AK001651.1 | 17 | 2147 | BAA91811.1 | |
| AK093570.1 | 11 | 1990 | ||
| AL137599.1 | 14 | 1650 | CAB70835.1 | |
| AY358572.1 | 17 | 2221 | AAQ88935.1 | |
| BC008337.1 | 17 | 2127 | AAH08337.1 | |
| BC015015.1 | 17 | 1986 | AAH15015.1 | |
| BC031550.1 | 17 | 2018 | AAH31550.1 |
More Information
Set Membership: |
|

Genomic Map