Assay Details
Target Gene Details
Entrez Gene ID: | 10899 |
Gene Name: | jumping translocation breakpoint |
Gene Aliases: |
HJTB, HSPC222, PAR, hJT |
Location: |
Chr.1:153974269-153977975 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JTB | NM_006694.3 | 5 | 1066 | NP_006685.1 |
| AB016488.1 | 5 | 775 | BAA33731.1 | |
| AF115850.2 | 5 | 764 | AAD09822.2 | |
| AF131797.1 | 5 | 647 | AAD20045.1 | |
| AF151056.1 | 4 | 822 | AAF36142.1 | |
| AK312106.1 | 5 | 748 | ||
| BC000499.1 | 5 | 788 | AAH00499.1 | |
| BC000996.2 | 5 | 715 | AAH00996.1 | |
| BC001363.1 | 5 | 753 | AAH01363.1 | |
| BC001667.1 | 5 | 718 | AAH01667.1 | |
| BC004239.1 | 5 | 775 | AAH04239.1 | |
| BC019277.1 | 5 | 709 | AAH19277.1 | |
| BT007285.1 | 5 | 343 | AAP35949.1 | |
| CR456985.1 | 5 | 343 | CAG33266.1 |
More Information
Set Membership: |
|

Genomic Map