Assay Details
Target Gene Details
Entrez Gene ID: | 55212 |
Gene Name: | Bardet-Biedl syndrome 7 |
Gene Aliases: |
BBS2L1 |
Location: |
Chr.4:121824329-121870520 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS7 | NM_176824.2 | 19 | 3284 | NP_789794.1 |
| XM_005263106.3 | 19 | 3310 | XP_005263163.1 | |
| XM_011532079.2 | 19 | 3355 | XP_011530381.1 | |
| XM_011532080.2 | 19 | 3352 | XP_011530382.1 | |
| XM_011532081.2 | 18 | 3190 | XP_011530383.1 | |
| XM_017008357.1 | 18 | 3142 | XP_016863846.1 | |
| XM_017008358.1 | 17 | 5297 | XP_016863847.1 | |
| BF431967.1 | 1 | 149 | ||
| BM998707.1 | 1 | 445 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map