Assay Details
Target Gene Details
Entrez Gene ID: | 84070 |
Gene Name: | family with sequence similarity 186 member B |
Gene Aliases: |
C12orf25 |
Location: |
Chr.12:49582875-49605710 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM186B | NM_032130.2 | 3 | 548 | NP_115506.1 |
| NR_027450.1 | 3 | 705 | ||
| XM_006719625.2 | 3 | 537 | XP_006719688.1 | |
| XM_006719626.2 | 3 | 537 | XP_006719689.1 | |
| XM_006719627.3 | 3 | 537 | XP_006719690.1 | |
| XM_011538796.2 | 3 | 537 | XP_011537098.1 | |
| XM_011538797.2 | 3 | 537 | XP_011537099.1 | |
| XM_017020008.1 | 3 | 788 | XP_016875497.1 | |
| AK093242.1 | 3 | 537 | ||
| AK302702.1 | 3 | 705 | ||
| AL136748.1 | 3 | 469 | CAB66682.1 | |
| BC035621.1 | 3 | 506 | AAH35621.1 | |
| DB087952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832406 | Chr.12:49466910 - 49668353 on Build GRCh38 | Loss |
|

Genomic Map