Assay Details
Target Gene Details
Entrez Gene ID: | 400794 |
Gene Name: | uncharacterized LOC400794 |
Gene Aliases: |
- |
Location: |
Chr.1:165476842-165582155 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC400794 | NR_026744.2 | |||
| AK093132.1 |
Target Gene Details
Entrez Gene ID: | 440699 |
Gene Name: | leucine rich repeat containing 52 |
Gene Aliases: |
- |
Location: |
Chr.1:165544241-165563948 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC52 | NM_001005214.3 | 2 | 777 | NP_001005214.2 |
| AA725678.1 | 1 | 349 | ||
| AK098677.1 | 2 | 1013 | BAC05375.1 | |
| BC132981.1 | 2 | 749 | ||
| BC132983.1 | 2 | 749 |
More Information
Set Membership: |
|

Genomic Map