Assay Details
Target Gene Details
Entrez Gene ID: | 9477 |
Gene Name: | mediator complex subunit 20 |
Gene Aliases: |
PRO0213, TRFP |
Location: |
Chr.6:41905353-41921147 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED20 | NM_001305455.1 | 3 | 1849 | NP_001292384.1 |
| NM_001305456.1 | 5 | 2121 | NP_001292385.1 | |
| NM_001305457.1 | 4 | 1928 | NP_001292386.1 | |
| NM_004275.4 | 4 | 2004 | NP_004266.2 | |
| NR_131160.1 | 3 | 1750 | ||
| AJ420544.1 | 3 | 1807 | ||
| AK023092.1 | 4 | 1948 | BAB14399.1 | |
| AK097385.1 | 5 | 2121 | ||
| AL050196.1 | 2 | 1519 | CAB43314.1 | |
| BC012618.1 | 4 | 2000 | AAH12618.1 | |
| BC019866.1 | 4 | 1981 | ||
| BC032552.1 | 4 | 1966 | AAH32552.1 |
More Information
Set Membership: |
|

Genomic Map