Assay Details
Target Gene Details
Entrez Gene ID: | 387775 |
Gene Name: | solute carrier family 22 member 10 |
Gene Aliases: |
OAT5, hOAT5 |
Location: |
Chr.11:63268257-63311983 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A10 | NM_001039752.3 | NP_001034841.3 | ||
| NR_134874.1 | ||||
| XM_011545016.2 | XP_011543318.1 | |||
| XM_017017699.1 | XP_016873188.1 | |||
| XM_017017700.1 | XP_016873189.1 | |||
| XM_017017701.1 | XP_016873190.1 | |||
| XM_017017702.1 | XP_016873191.1 | |||
| XM_017017703.1 | XP_016873192.1 | |||
| XM_017017704.1 | XP_016873193.1 | |||
| AB075876.1 | BAD38658.1 | |||
| AK296299.1 | ||||
| AK298430.1 | ||||
| BX091233.1 | ||||
| DR003989.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759833 | Chr.11:63277520 - 63359382 on Build GRCh38 | Loss |
|
| nsv555177 | Chr.11:63080046 - 63343097 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map