Assay Details
Target Gene Details
Entrez Gene ID: | 683 |
Gene Name: | bone marrow stromal cell antigen 1 |
Gene Aliases: |
CD157 |
Location: |
Chr.4:15686347-15774177 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BST1 | NM_004334.2 | NP_004325.2 | ||
| XM_005248185.1 | XP_005248242.1 | |||
| XM_005248186.1 | XP_005248243.1 | |||
| XM_011513878.2 | XP_011512180.1 | |||
| XM_011513879.1 | XP_011512181.1 | |||
| XM_011513881.2 | XP_011512183.1 | |||
| XM_017008565.1 | XP_016864054.1 | |||
| XM_017008566.1 | XP_016864055.1 | |||
| XM_017008567.1 | XP_016864056.1 | |||
| AB175627.1 | BAE92566.1 | |||
| AK312497.1 | ||||
| BC012095.1 | AAH12095.1 | |||
| BT019502.1 | AAV38309.1 | |||
| BU732655.1 | ||||
| D21878.1 | BAA04885.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10459 | Chr.4:15719720 - 15722959 on Build GRCh38 | Loss |
|
| nsv428439 | Chr.4:15555338 - 15747897 on Build GRCh38 | Gain+Loss |
|
| esv2759227 | Chr.4:15442993 - 15747897 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map