Assay Details
Target Gene Details
Entrez Gene ID: | 79157 |
Gene Name: | major facilitator superfamily domain containing 11 |
Gene Aliases: |
ET |
Location: |
Chr.17:76736565-76803805 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD11 | NM_001242534.1 | 1 | 331 | NP_001229463.1 |
| AF015186.1 | 1 | 43 | AAB92497.1 | |
| AF015187.1 | 1 | 43 | ||
| AF015188.1 | 2 | 870 | ||
| AF015189.1 | 1 | 585 | ||
| AF015190.1 | 1 | 331 |
Target Gene Details
Entrez Gene ID: | 6427 |
Gene Name: | serine and arginine rich splicing factor 2 |
Gene Aliases: |
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b |
Location: |
Chr.17:76734115-76737431 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF2 | NM_001195427.1 | 1 | 517 | NP_001182356.1 |
| NM_003016.4 | 1 | 517 | NP_003007.2 | |
| NR_036608.1 | 1 | 517 | ||
| XM_017024942.1 | 1 | 537 | XP_016880431.1 | |
| AK092489.1 | BAC03903.1 | |||
| AK093626.1 | BAC04206.1 | |||
| AK097069.1 | ||||
| AK098209.1 | 2 | 408 | ||
| AK123712.1 | ||||
| AK124792.1 | 1 | 480 | ||
| AK126432.1 | ||||
| AK223252.1 | 1 | 436 | BAD96972.1 | |
| AK297813.1 | 1 | 437 | ||
| BC000339.2 | 1 | 417 | AAH00339.1 | |
| BC001303.2 | 1 | 414 | AAH01303.1 | |
| BC066958.1 | AAH66958.1 | |||
| BC070086.1 | 1 | 437 | AAH70086.1 | |
| BT007250.1 | 1 | 266 | AAP35914.1 | |
| DA513281.1 | 1 | 485 | ||
| DC361254.1 | 1 | 517 | ||
| M90104.1 | 1 | 421 | AAA60306.1 | |
| X62447.1 | 1 | 302 | CAA44307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
| nsv828110 | Chr.17:76736727 - 76737636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map