Assay Details
Target Gene Details
Entrez Gene ID: | 56267 |
Gene Name: | kynurenine aminotransferase 3 |
Gene Aliases: |
CCBL2, KAT3, KATIII |
Location: |
Chr.1:88935773-88992960 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KYAT3 | NM_001008661.2 | 2 | 432 | NP_001008661.1 |
| NM_001008662.2 | NP_001008662.1 | |||
| AK057176.1 | ||||
| AK301326.1 | 2 | 119 | ||
| AK315917.1 | 2 | 129 | ||
| AL833593.1 | 2 | 77 | ||
| AY028624.1 | 2 | 252 | AAK26163.1 | |
| BM728982.1 | 2 | 394 | ||
| CR627392.1 | 2 | 94 | CAH10487.1 | |
| DC424120.1 | 2 | 435 |
Target Gene Details
Entrez Gene ID: | 494115 |
Gene Name: | RNA binding motif protein, X-linked like 1 |
Gene Aliases: |
RBM1 |
Location: |
Chr.1:88979456-88992960 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMXL1 | NM_001162536.2 | 2 | 432 | NP_001156008.1 |
| NM_019610.5 | NP_062556.2 | |||
| AL832554.1 | CAI46148.1 | |||
| BC012942.1 | AAH12942.1 | |||
| DA010727.1 | ||||
| DA216732.1 | 2 | 274 | ||
| DA390224.1 | ||||
| DC397531.1 | 2 | 499 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv207n100 | Chr.1:88932165 - 88995971 on Build GRCh38 | Loss |
|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
| nsv1002798 | Chr.1:88954378 - 88988906 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map