Assay Details
Target Gene Details
Entrez Gene ID: | 64063 |
Gene Name: | protease, serine 22 |
Gene Aliases: |
BSSP-4, SP001LA, hBSSP-4 |
Location: |
Chr.16:2852727-2858630 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS22 | NM_022119.3 | NP_071402.1 | ||
| XM_005255473.3 | XP_005255530.1 | |||
| AB010779.1 | BAB20263.1 | |||
| AF321182.1 | AAG35070.1 | |||
| AW083723.1 | ||||
| AY358396.1 | AAQ88762.1 | |||
| BC009726.1 | AAH09726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1712 | Chr.16:2842984 - 2870374 on Build GRCh38 | Insertion |
|
| nsv952905 | Chr.16:2817200 - 2863199 on Build GRCh38 | Deletion |
|
| nsv509587 | Chr.16:2842958 - 2874324 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map