Assay Details
Target Gene Details
| Entrez Gene ID: | 90527 | 
| Gene Name: | dual oxidase maturation factor 1 | 
| Gene Aliases: | NIP, NUMBIP, mol | 
| Location: | Chr.15:45117366-45129938 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 9 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DUOXA1 | NM_001276264.1 | NP_001263193.1 | ||
| NM_001276265.1 | NP_001263194.1 | |||
| NM_001276267.1 | 9 | 2554 | NP_001263196.1 | |
| NM_001276268.1 | 6 | 2214 | NP_001263197.1 | |
| NM_144565.3 | NP_653166.2 | |||
| XM_006720743.3 | 9 | 2884 | XP_006720806.1 | |
| XM_006720745.1 | 9 | 2859 | XP_006720808.1 | |
| XM_006720746.1 | 9 | 2888 | XP_006720809.1 | |
| XM_006720747.3 | 8 | 2796 | XP_006720810.1 | |
| XM_006720749.1 | 9 | 2654 | XP_006720812.1 | |
| XM_006720751.3 | 7 | 2377 | XP_006720814.1 | |
| XM_006720752.3 | 7 | 2386 | XP_006720815.2 | |
| XM_006720754.1 | 8 | 2519 | XP_006720817.1 | |
| XM_006720755.1 | 8 | 2495 | XP_006720818.1 | |
| XM_011522181.2 | 9 | 2912 | XP_011520483.1 | |
| XM_011522182.1 | 8 | 2627 | XP_011520484.1 | |
| XM_011522183.2 | 10 | 2742 | XP_011520485.1 | |
| AK056896.1 | 6 | 2214 | BAB71304.1 | |
| AK292771.1 | ||||
| BC029819.1 | AAH29819.1 | |||
| CB850544.1 | 1 | 496 | ||
| EU927396.1 | ||||
| EU927397.1 | 
Target Gene Details
| Entrez Gene ID: | 405753 | 
| Gene Name: | dual oxidase maturation factor 2 | 
| Gene Aliases: | SIMNIPHOM, TDH5 | 
| Location: | Chr.15:45114321-45118106 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DUOXA2 | NM_207581.3 | 6 | 1179 | NP_997464.2 | 
| XM_017022180.1 | 6 | 1234 | XP_016877669.1 | |
| BC137465.1 | 6 | 1100 | ||
| BM987789.1 | ||||
| BX537581.1 | 6 | 1463 | ||
| DQ489734.1 | 6 | 1030 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv569312 | Chr.15:45089800 - 45168224 on Build GRCh38 | Loss |  DUOX2  DUOX1  DUOXA1  SHF  DUOXA2 | 
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |  SORD  DUOX2  B2M  DUOX1  LOC100419583  DUOXA1  PATL2  TERB2  TRIM69  DUOXA2 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top