Assay Details
Target Gene Details
Entrez Gene ID: | 56916 |
Gene Name: | SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1 |
Gene Aliases: |
ADERM, ETL1, HEL1 |
Location: |
Chr.4:94207608-94291292 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCAD1 | NM_001128429.2 | NP_001121901.1 | ||
| NM_001128430.1 | NP_001121902.1 | |||
| NM_020159.4 | NP_064544.2 | |||
| NR_045644.1 | ||||
| XM_017008463.1 | XP_016863952.1 | |||
| XM_017008464.1 | XP_016863953.1 | |||
| XM_017008465.1 | XP_016863954.1 | |||
| AB032948.2 | ||||
| AK025302.1 | ||||
| AK027490.1 | BAB55150.1 | |||
| AY008271.1 | AAG16639.1 | |||
| BC017953.1 | ||||
| BC045534.1 | AAH45534.1 |
More Information
Set Membership: |
|

Genomic Map