Assay Details
Target Gene Details
Entrez Gene ID: | 5000 |
Gene Name: | origin recognition complex subunit 4 |
Gene Aliases: |
ORC4L, ORC4P |
Location: |
Chr.2:147930397-148021604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORC4 | NM_001190879.2 | NP_001177808.1 | ||
| NM_001190881.2 | NP_001177810.1 | |||
| NM_001190882.2 | NP_001177811.1 | |||
| NM_002552.4 | NP_002543.2 | |||
| NM_181741.3 | NP_859525.1 | |||
| NM_181742.3 | NP_859526.1 | |||
| XM_011511255.2 | XP_011509557.1 | |||
| XM_017004226.1 | XP_016859715.1 | |||
| XM_017004227.1 | XP_016859716.1 | |||
| AF022108.1 | AAC01957.1 | |||
| AF047598.1 | AAC80282.1 | |||
| AF132596.1 | AAD22110.1 | |||
| AK055257.1 | 1 | 1673 | ||
| AK225092.1 | ||||
| AK291989.1 | ||||
| AK295721.1 | ||||
| AK298862.1 | ||||
| AK299749.1 | ||||
| BC014847.1 | AAH14847.1 | |||
| BC016177.1 | AAH16177.1 |
More Information
Set Membership: |
|

Genomic Map