Assay Details
Target Gene Details
Entrez Gene ID: | 7456 |
Gene Name: | WAS/WASL interacting protein family member 1 |
Gene Aliases: |
PRPL-2, WAS2, WASPIP, WIP |
Location: |
Chr.2:174559574-174682899 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WIPF1 | NM_001077269.1 | 8 | 3534 | NP_001070737.1 |
| NM_003387.4 | 8 | 3470 | NP_003378.3 | |
| XM_011511780.2 | 8 | 3656 | XP_011510082.1 | |
| AK097901.1 | 1 | 1129 | ||
| AL832275.1 | 1 | 1538 | ||
| BG820214.1 | 1 | 206 | ||
| BM475558.1 | 1 | 64 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3041 | Chr.2:174556013 - 174576388 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map