Assay Details
Target Gene Details
Entrez Gene ID: | 5621 |
Gene Name: | prion protein |
Gene Aliases: |
ASCR, AltPrP, CD230, CJD, GSS, KURU, PRIP, PrP, PrP27-30, PrP33-35C, PrPc, p27-30 |
Location: |
Chr.20:4686151-4701588 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRNP | NM_000311.3 | 2 | 1958 | NP_000302.1 |
| NM_001080121.1 | 2 | 1953 | NP_001073590.1 | |
| NM_001080122.1 | 2 | 1949 | NP_001073591.1 | |
| NM_001080123.1 | 2 | 1822 | NP_001073592.1 | |
| NM_001271561.1 | 2 | 1958 | NP_001258490.1 | |
| NM_183079.2 | 2 | 1954 | NP_898902.1 | |
| AK090575.1 | 3 | 1557 | ||
| AY008282.1 | 2 | 1629 | AAG21693.1 | |
| BC012844.1 | 2 | 1633 | AAH12844.1 | |
| BC022532.1 | 2 | 1646 | AAH22532.1 | |
| D00015.1 | 1 | 1641 | BAA00011.1 | |
| M13667.1 | 1 | 1641 | AAA19664.1 | |
| M13899.1 | 2 | 1635 | AAA60182.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058830 | Chr.20:4663154 - 4725219 on Build GRCh38 | Gain |
|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
| nsv585310 | Chr.20:4586645 - 4794039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map