Assay Details
Target Gene Details
Entrez Gene ID: | 50810 |
Gene Name: | hepatoma-derived growth factor, related protein 3 |
Gene Aliases: |
CGI-142, HDGF-2, HDGF2, HRP-3 |
Location: |
Chr.15:83112738-83208018 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDGFRP3 | NM_016073.3 | 6 | 1673 | NP_057157.1 |
| XM_006720554.3 | XP_006720617.1 | |||
| AB029156.1 | 6 | 1189 | BAA90477.1 | |
| AF110642.1 | 6 | 1193 | AAM27001.1 | |
| AF151900.1 | 6 | 1193 | AAD34137.1 | |
| AK001280.1 | 6 | 1224 | BAA91597.1 | |
| AL109689.1 | 2 | 590 | CAB52021.1 | |
| BC015483.1 | 6 | 1167 | AAH15483.1 |
Target Gene Details
Entrez Gene ID: | 53346 |
Gene Name: | transmembrane 6 superfamily member 1 |
Gene Aliases: |
- |
Location: |
Chr.15:83107572-83145403 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM6SF1 | XM_017022322.1 | XP_016877811.1 | ||
| XM_017022331.1 | XP_016877820.1 |
More Information
Set Membership: |
|

Genomic Map