Assay Details
Target Gene Details
Entrez Gene ID: | 8854 |
Gene Name: | aldehyde dehydrogenase 1 family member A2 |
Gene Aliases: |
RALDH(II), RALDH2, RALDH2-T |
Location: |
Chr.15:57953424-58065923 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1A2 | NM_001206897.1 | 14 | 3452 | NP_001193826.1 |
| NM_003888.3 | 13 | 3355 | NP_003879.2 | |
| NM_170696.2 | 12 | 3241 | NP_733797.1 | |
| NM_170697.2 | 11 | 2959 | NP_733798.1 | |
| AA447978.1 | 1 | 252 | ||
| AB015228.1 | 11 | 2850 | BAA34787.1 | |
| AK128709.1 | 13 | 3850 | ||
| AL110274.1 | 1 | 4152 | ||
| AL110299.1 | 5 | 2903 | CAB53740.2 | |
| AL137418.1 | 1 | 1287 | ||
| BC030589.2 | 12 | 2999 | AAH30589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569593 | Chr.15:57909500 - 58016452 on Build GRCh38 | Loss |
|
| nsv833023 | Chr.15:57869641 - 58047976 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map