Assay Details
Target Gene Details
Entrez Gene ID: | 7042 |
Gene Name: | transforming growth factor beta 2 |
Gene Aliases: |
LDS4, TGF-beta2 |
Location: |
Chr.1:218345334-218444619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB2 | NM_001135599.2 | 1 | 1416 | NP_001129071.1 |
| NM_003238.3 | 1 | 1416 | NP_003229.1 | |
| AB209842.1 | 1 | 1344 | BAD93079.1 | |
| AK296504.1 | 1 | 49 | ||
| AK313106.1 | 1 | 153 | ||
| BC096235.4 | 1 | 202 | AAH96235.1 | |
| BC099635.1 | 1 | 202 | AAH99635.1 | |
| M19154.1 | 1 | 515 | AAA50404.1 | |
| Y00083.1 | 1 | 224 | CAA68279.1 |
More Information
Set Membership: |
|

Genomic Map