Assay Details
Target Gene Details
Entrez Gene ID: | 23498 |
Gene Name: | 3-hydroxyanthranilate 3,4-dioxygenase |
Gene Aliases: |
3-HAO, HAO |
Location: |
Chr.2:42766198-42794359 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAAO | NM_012205.2 | NP_036337.2 | ||
| XM_005264230.3 | XP_005264287.1 | |||
| XM_011532729.2 | XP_011531031.1 | |||
| XM_011532730.2 | XP_011531032.1 | |||
| XM_011532731.2 | XP_011531033.1 | |||
| XM_017003717.1 | XP_016859206.1 | |||
| BC029510.1 | AAH29510.1 | |||
| BI602406.1 | ||||
| CB850657.1 | ||||
| CR457063.1 | CAG33344.1 | |||
| Z29481.1 | CAA82618.1 |
Target Gene Details
Entrez Gene ID: | 57504 |
Gene Name: | metastasis associated 1 family member 3 |
Gene Aliases: |
- |
Location: |
Chr.2:42494569-42795032 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTA3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519877 | Chr.2:42751296 - 42793774 on Build GRCh38 | Loss |
|
| nsv457152 | Chr.2:42752929 - 42787149 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map