Assay Details
Target Gene Details
Entrez Gene ID: | 9373 |
Gene Name: | phospholipase A2 activating protein |
Gene Aliases: |
DOA1, PLA2P, PLAP |
Location: |
Chr.9:26903370-26947473 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLAA | NM_001031689.2 | 14 | 2916 | NP_001026859.1 |
| NM_001321546.1 | 13 | 2850 | NP_001308475.1 | |
| AF145020.1 | 14 | 2715 | AAD42075.1 | |
| AJ238243.1 | 13 | 2348 | CAB42881.1 | |
| AK001642.1 | 14 | 2619 | BAA91803.1 | |
| AL133608.1 | 14 | 2909 | CAB63739.1 | |
| BC032551.1 | 14 | 2598 | AAH32551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528614 | Chr.9:26893664 - 27074235 on Build GRCh38 | Gain |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map