Assay Details
Target Gene Details
Entrez Gene ID: | 8888 |
Gene Name: | minichromosome maintenance complex component 3 associated protein |
Gene Aliases: |
GANP, MAP80, SAC3 |
Location: |
Chr.21:46235125-46286274 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3AP | NM_003906.4 | 27 | 5810 | NP_003897.2 |
| XM_005261203.4 | 28 | 6386 | XP_005261260.1 | |
| XM_005261204.4 | 28 | 6282 | XP_005261261.1 | |
| XM_005261205.3 | 28 | 5898 | XP_005261262.1 | |
| AB005543.1 | 12 | 2295 | BAA25170.1 | |
| AB011144.1 | 27 | 5380 | BAA25498.1 | |
| AJ010089.1 | 27 | 5739 | CAB52687.1 | |
| AK026578.1 | 6 | 1153 | ||
| AK095181.1 | 7 | 2763 | ||
| AK126313.1 | 17 | 4215 | ||
| AK127239.1 | 17 | 3699 | ||
| AK304208.1 | 27 | 4669 | ||
| BC004497.2 | 8 | 1460 | AAH04497.2 | |
| BC013285.2 | 12 | 2096 | AAH13285.2 | |
| BC104958.1 | 27 | 5778 | AAI04959.1 | |
| BC104960.1 | 27 | 5778 | AAI04961.1 |
Target Gene Details
Entrez Gene ID: | 114044 |
Gene Name: | MCM3AP antisense RNA 1 |
Gene Aliases: |
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031 |
Location: |
Chr.21:46229231-46251701 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3AP-AS1 | NR_002776.4 | |||
| NR_110565.1 | ||||
| NR_110566.1 | ||||
| NR_110567.1 | ||||
| AF426263.1 | ||||
| AK001370.1 | ||||
| BC059792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|
| esv2641041 | Chr.21:46236589 - 46239240 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map