Assay Details
Target Gene Details
Entrez Gene ID: | 10060 |
Gene Name: | ATP binding cassette subfamily C member 9 |
Gene Aliases: |
ABC37, ATFB12, CANTU, CMD1O, SUR2 |
Location: |
Chr.12:21797389-21941869 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCC9 | NM_020297.3 | 38 | 6059 | NP_064693.2 |
| XM_005253284.3 | 40 | 6845 | XP_005253341.1 | |
| XM_005253286.3 | 41 | 6905 | XP_005253343.1 | |
| XM_005253287.4 | 41 | 7021 | XP_005253344.1 | |
| XM_005253288.3 | 39 | 6729 | XP_005253345.1 | |
| XM_005253289.3 | 39 | 6806 | XP_005253346.1 | |
| XM_005253290.3 | 38 | 6704 | XP_005253347.1 | |
| XM_006719025.3 | 40 | 6982 | XP_006719088.1 | |
| XM_011520545.2 | 40 | 6901 | XP_011518847.1 | |
| AK092535.1 | ||||
| BX537513.1 | 1 | 1303 | ||
| BX640893.1 | 1 | 994 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv557744 | Chr.12:21535966 - 21813767 on Build GRCh38 | Loss |
|
| esv3580185 | Chr.12:21780932 - 21799634 on Build GRCh38 | Loss |
|
| nsv470275 | Chr.12:21785610 - 21973380 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map