Assay Details
Target Gene Details
Entrez Gene ID: | 5564 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit beta 1 |
Gene Aliases: |
AMPK, HAMPKb |
Location: |
Chr.12:119667864-119681624 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAB1 | NM_006253.4 | 1 | 267 | NP_006244.2 |
| XM_005253909.1 | XP_005253966.1 | |||
| AF022116.1 | 1 | 246 | AAC98897.1 | |
| AJ224515.1 | CAA12024.1 | |||
| AK301165.1 | ||||
| AK312495.1 | ||||
| BC001007.2 | 1 | 244 | AAH01007.1 | |
| BC001056.2 | 1 | 244 | AAH01056.1 | |
| BC001823.2 | 1 | 244 | AAH01823.1 | |
| BC017671.1 | 1 | 253 | AAH17671.1 | |
| BU539177.1 | 1 | 268 | ||
| U83994.1 | 1 | 223 | AAD09237.1 | |
| Y12556.1 | 1 | 59 | CAA73146.1 |
More Information
Set Membership: |
|

Genomic Map