Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D3 | NM_001015881.1 | 3 | 848 | NP_001015881.1 |
| NM_001318468.1 | 4 | 1316 | NP_001305397.1 | |
| NM_001318470.1 | 4 | 1302 | NP_001305399.1 | |
| NM_004089.3 | 3 | 1053 | NP_004080.2 | |
| NM_198057.2 | 3 | 1369 | NP_932174.1 | |
| XM_005262099.1 | 5 | 1330 | XP_005262156.1 | |
| XM_005262100.1 | 4 | 1205 | XP_005262157.1 | |
| XM_005262102.1 | 4 | 1198 | XP_005262159.1 | |
| XM_005262103.3 | 4 | 1308 | XP_005262160.1 | |
| XM_011530884.1 | 4 | 1404 | XP_011529186.1 | |
| XM_017029335.1 | 4 | 1140 | XP_016884824.1 | |
| AB025432.1 | 3 | 1036 | BAB18680.1 | |
| AF153603.1 | 3 | 1031 | AAD41085.1 | |
| AF183393.1 | 3 | 1039 | AAD56234.1 | |
| AF228339.1 | 3 | 1043 | AAG12456.1 | |
| AK092645.1 | 4 | 1255 | BAC03934.1 | |
| AK092669.1 | 3 | 1341 | ||
| AK127938.1 | 3 | 993 | ||
| AK300535.1 | 4 | 1180 | ||
| AL110191.1 | 3 | 1052 | CAB53669.1 | |
| AY007119.1 | 3 | 952 | ||
| BC018148.1 | 3 | 735 | ||
| BC072446.1 | 3 | 1047 | AAH72446.1 | |
| BX647854.1 | 4 | 1302 | ||
| CR933650.1 | 3 | 1055 | CAI45951.1 |
More Information
Set Membership: |
|

Genomic Map