Assay Details
Target Gene Details
Entrez Gene ID: | 255626 |
Gene Name: | histone cluster 1, H2ba |
Gene Aliases: |
H2BFU, STBP, TH2B, TSH2B, TSH2B.1, bA317E16.3 |
Location: |
Chr.6:25726909-25727345 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2BA | NM_170610.2 | NP_733759.1 | ||
| BC066238.1 | AAH66238.1 | |||
| BC066239.1 | AAH66239.1 | |||
| BC066240.1 | AAH66240.1 | |||
| BC066241.1 | AAH66241.1 | |||
| BC066242.1 | AAH66242.1 | |||
| BC066243.1 | AAH66243.1 |
Target Gene Details
Entrez Gene ID: | 6568 |
Gene Name: | solute carrier family 17 member 1 |
Gene Aliases: |
NAPI-1, NPT-1, NPT1 |
Location: |
Chr.6:25723744-25832108 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A1 | XM_017011201.1 | XP_016866690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3570813 | Chr.6:25724901 - 25728340 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map