Assay Details
Target Gene Details
| Entrez Gene ID: | 378708 | 
| Gene Name: | apoptosis-inducing, TAF9-like domain 1 | 
| Gene Aliases: | CENP-S, CENPS, FAAP16, MHF1 | 
| Location: | Chr.1:10430102-10442815 on Build GRCh38 | 
| Assay Gene Location: | Overlaps - Exon 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| APITD1 | NM_199294.2 | NP_954988.1 | ||
| NR_036462.1 | ||||
| BE858209.1 | ||||
| BG213105.1 | ||||
| BU902753.1 | 
Target Gene Details
| Entrez Gene ID: | 100526739 | 
| Gene Name: | APITD1-CORT readthrough | 
| Gene Aliases: | - | 
| Location: | Chr.1:10430102-10452003 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| APITD1-CORT | NM_001243768.1 | NP_001230697.1 | ||
| NM_001270517.1 | NP_001257446.1 | |||
| NM_198544.3 | NP_940946.1 | |||
| NM_199006.2 | NP_950171.2 | |||
| NR_037187.1 | ||||
| AA858169.1 | ||||
| AF516753.1 | AAM61954.1 | |||
| AF521016.1 | AAM70482.1 | |||
| CF128105.1 | ||||
| HY054077.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2764192 | Chr.1:10323676 - 10534129 on Build GRCh38 | Gain |  CORT  APITD1  PGD  KIF1B  APITD1-CORT  PEX14  DFFA | 
| esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |  CORT  APITD1  PGD  KIF1B  APITD1-CORT  MIR1273D  PEX14  DFFA  UBE4B | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top