Assay Details
Target Gene Details
Entrez Gene ID: | 23216 |
Gene Name: | TBC1 domain family member 1 |
Gene Aliases: |
TBC, TBC1 |
Location: |
Chr.4:37891084-38139175 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D1 | NM_001253912.1 | 2 | 378 | NP_001240841.1 |
| NM_015173.3 | 2 | 378 | NP_055988.2 | |
| XM_005262646.2 | 2 | 378 | XP_005262703.1 | |
| XM_005262647.2 | 2 | 378 | XP_005262704.1 | |
| XM_011513659.2 | 2 | 378 | XP_011511961.1 | |
| XM_011513660.2 | 2 | 378 | XP_011511962.1 | |
| XM_011513662.2 | 2 | 378 | XP_011511964.1 | |
| XM_011513663.2 | 2 | 378 | XP_011511965.1 | |
| XM_011513664.2 | 2 | 378 | XP_011511966.1 | |
| XM_011513665.2 | 2 | 378 | XP_011511967.1 | |
| XM_011513666.2 | 2 | 378 | XP_011511968.1 | |
| XM_011513668.2 | 2 | 378 | XP_011511970.1 | |
| XM_017007918.1 | 2 | 378 | XP_016863407.1 | |
| XM_017007919.1 | 2 | 378 | XP_016863408.1 | |
| XM_017007920.1 | 2 | 378 | XP_016863409.1 | |
| XM_017007922.1 | 2 | 378 | XP_016863411.1 | |
| AB449881.1 | ||||
| AK295746.1 | 2 | 201 | ||
| BC050321.1 | 2 | 375 | AAH50321.3 | |
| BC126979.1 | 1 | 122 | ||
| DB036208.1 | 2 | 378 |
More Information
Set Membership: |
|

Genomic Map