Assay Details
Target Gene Details
Entrez Gene ID: | 92737 |
Gene Name: | delta/notch like EGF repeat containing |
Gene Aliases: |
UNQ26, bet |
Location: |
Chr.2:229357629-229714570 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNER | NM_139072.3 | NP_620711.3 | ||
| XM_005246950.2 | XP_005247007.1 | |||
| AF442487.1 | AAM21557.1 | |||
| AK289553.1 | ||||
| AK304596.1 | ||||
| AK304631.1 | ||||
| AK308576.1 | ||||
| AL137311.1 | CAB70690.1 | |||
| AY358891.1 | AAQ89250.1 | |||
| BC024766.2 | AAH24766.2 | |||
| BC035009.1 | AAH35009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834564 | Chr.2:229239150 - 229431420 on Build GRCh38 | Gain |
|
| nsv3198 | Chr.2:229341777 - 229372836 on Build GRCh38 | Insertion |
|
| esv3594572 | Chr.2:229347352 - 229417034 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map