Assay Details
Target Gene Details
Entrez Gene ID: | 10349 |
Gene Name: | ATP binding cassette subfamily A member 10 |
Gene Aliases: |
EST698739 |
Location: |
Chr.17:69148007-69244815 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 36 - Exon 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA10 | NM_080282.3 | NP_525021.3 | ||
| AK298508.1 | ||||
| AL832004.2 | CAD89903.1 | |||
| AY028900.1 | AAK30025.1 | |||
| AY247065.1 | AAO72161.1 | |||
| BC031026.1 | AAH31026.1 | |||
| BC051320.1 | AAH51320.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2128 | Chr.17:69134289 - 69168771 on Build GRCh38 | Insertion |
|
| nsv470598 | Chr.17:69085689 - 69216454 on Build GRCh38 | Loss |
|
| nsv575930 | Chr.17:69123884 - 69215890 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map