Assay Details
Target Gene Details
Entrez Gene ID: | 343472 |
Gene Name: | BarH like homeobox 2 |
Gene Aliases: |
- |
Location: |
Chr.1:90712022-90717237 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BARHL2 | NM_020063.1 | NP_064447.1 | ||
| AY338397.1 | AAQ01189.1 | |||
| BC126439.1 | ||||
| BC126441.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950204 | Chr.1:90706244 - 90719643 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map