Assay Details
Target Gene Details
Entrez Gene ID: | 4830 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 1 |
Gene Aliases: |
AWD, GAAD, NB, NBS, NDKA, NDPK-A, NDPKA, NM23, NM23-H1 |
Location: |
Chr.17:51153559-51162089 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME1 | NM_000269.2 | NP_000260.1 | ||
| NM_198175.1 | NP_937818.1 | |||
| AF487339.1 | AAO85436.1 | |||
| AK291105.1 | ||||
| AL360166.1 | ||||
| AL360191.1 | ||||
| BC000293.2 | AAH00293.1 | |||
| BC018994.2 | AAH18994.1 | |||
| BE727815.1 | ||||
| BF965472.1 | ||||
| BG531372.1 | ||||
| BG715560.1 | ||||
| BG757842.1 | ||||
| BI666785.1 | ||||
| BU600979.1 | ||||
| CR542104.1 | CAG46901.1 | |||
| CR542115.1 | CAG46912.1 | |||
| X17620.1 | CAA35621.1 | |||
| X73066.1 | CAA51527.1 |
Target Gene Details
Entrez Gene ID: | 654364 |
Gene Name: | NME1-NME2 readthrough |
Gene Aliases: |
NM23-LV, NMELV |
Location: |
Chr.17:51153536-51171744 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME1-NME2 | NM_001018136.2 | NP_001018146.1 | ||
| NR_037149.1 | ||||
| AK313448.1 | ||||
| BC107894.1 | AAI07895.1 | |||
| BC133029.1 | ||||
| BC133031.1 | ||||
| BI603164.1 | ||||
| DQ109675.1 | AAZ82097.1 |
More Information
Set Membership: |
|

Genomic Map