Assay Details
Target Gene Details
Entrez Gene ID: | 23431 |
Gene Name: | adaptor related protein complex 4 epsilon 1 subunit |
Gene Aliases: |
CPSQ4, SPG51, STUT1 |
Location: |
Chr.15:50908569-51005900 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4E1 | NM_001252127.1 | 7 | 910 | NP_001239056.1 |
| NM_007347.4 | 7 | 887 | NP_031373.2 | |
| XM_005254264.3 | 7 | 705 | XP_005254321.1 | |
| XM_006720447.3 | 7 | 1065 | XP_006720510.1 | |
| XM_017022042.1 | 7 | 832 | XP_016877531.1 | |
| AB030653.1 | 7 | 810 | BAA82969.1 | |
| AF155156.2 | 7 | 819 | AAD43326.2 | |
| AK297290.1 | 7 | 887 | ||
| BC126308.1 | 7 | 886 | ||
| BC130466.1 | 7 | 886 | ||
| CR749604.1 | 7 | 787 | CAH18399.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569403 | Chr.15:50899224 - 51012176 on Build GRCh38 | Loss |
|
| nsv569398 | Chr.15:50496928 - 51002798 on Build GRCh38 | Loss |
|
| nsv1047443 | Chr.15:50452584 - 51354826 on Build GRCh38 | Gain |
|
| nsv1038778 | Chr.15:50904444 - 51064531 on Build GRCh38 | Loss |
|
| dgv4526n54 | Chr.15:50582767 - 51475068 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map