Assay Details
Target Gene Details
Entrez Gene ID: | 2673 |
Gene Name: | glutamine--fructose-6-phosphate transaminase 1 |
Gene Aliases: |
CMS12, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, MSLG |
Location: |
Chr.2:69319769-69387254 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFPT1 | NM_001244710.1 | 20 | 2678 | NP_001231639.1 |
| NM_002056.3 | 19 | 2624 | NP_002047.2 | |
| XM_017003801.1 | 20 | 2642 | XP_016859290.1 | |
| XM_017003802.1 | 19 | 2580 | XP_016859291.1 | |
| BC045641.1 | 19 | 2620 | AAH45641.1 | |
| M90516.1 | 19 | 2563 | AAA58502.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33940 | Chr.2:69115288 - 69789041 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map