Assay Details
Target Gene Details
Entrez Gene ID: | 390197 |
Gene Name: | olfactory receptor family 4 subfamily D member 10 |
Gene Aliases: |
OR11-251, OR4D10P, OST711 |
Location: |
Chr.11:59477430-59478365 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D10 | NM_001004705.1 | NP_001004705.1 | ||
| BC136893.1 | 1 | 957 | ||
| BC136894.1 | 1 | 957 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983027 | Chr.11:59477533 - 59479704 on Build GRCh38 | Duplication |
|
| esv3626537 | Chr.11:59478051 - 59492222 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map