Assay Details
Target Gene Details
Entrez Gene ID: | 54898 |
Gene Name: | ELOVL fatty acid elongase 2 |
Gene Aliases: |
SSC2 |
Location: |
Chr.6:10980759-11044391 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL2 | NM_017770.3 | 8 | 3940 | NP_060240.3 |
| XM_011514716.2 | 8 | 3892 | XP_011513018.1 | |
| AJ420504.1 | 1 | 1696 | ||
| AL080199.1 | 1 | 895 | ||
| BC050278.1 | 8 | 3940 | AAH50278.2 | |
| BC060809.1 | 8 | 3891 | AAH60809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661047 | Chr.6:10978724 - 10981096 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map