Assay Details
Target Gene Details
Entrez Gene ID: | 1655 |
Gene Name: | DEAD-box helicase 5 |
Gene Aliases: |
G17P1, HLR1, HUMP68, p68 |
Location: |
Chr.17:64498254-64507038 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX5 | NM_001320595.1 | 7 | 974 | NP_001307524.1 |
NM_001320596.1 | 7 | 863 | NP_001307525.1 | |
NM_001320597.1 | 6 | 714 | NP_001307526.1 | |
NM_004396.4 | 6 | 949 | NP_004387.1 | |
AB209919.1 | 6 | 701 | BAD93156.1 | |
AB451257.1 | 6 | 547 | ||
AB451382.1 | 6 | 547 | ||
AK297192.1 | 5 | 479 | ||
AK297753.1 | 7 | 684 | ||
AK297905.1 | 5 | 507 | ||
AK310318.1 | 6 | 593 | ||
BC016027.1 | 6 | 688 | AAH16027.1 | |
BT006943.1 | 6 | 547 | AAP35589.1 | |
BX571764.1 | 6 | 718 | CAE11890.1 | |
X15729.1 | 6 | 714 | CAA33751.1 | |
X52104.1 | 6 | 722 | CAA36324.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
