Assay Details
Target Gene Details
Entrez Gene ID: | 144608 |
Gene Name: | chromosome 12 open reading frame 60 |
Gene Aliases: |
- |
Location: |
Chr.12:14567991-14823858 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C12orf60 | XM_017018871.1 | XP_016874360.1 | ||
| XM_017018872.1 | XP_016874361.1 |
Target Gene Details
Entrez Gene ID: | 121504 |
Gene Name: | histone cluster 4, H4 |
Gene Aliases: |
H4/p |
Location: |
Chr.12:14770720-14771131 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST4H4 | NM_175054.2 | 1 | 64 | NP_778224.1 |
| AK311916.1 | 1 | 64 |
More Information
Set Membership: |
|

Genomic Map