Assay Details
Target Gene Details
Entrez Gene ID: | 10717 |
Gene Name: | adaptor related protein complex 4 beta 1 subunit |
Gene Aliases: |
BETA-4, CPSQ5, SPG47 |
Location: |
Chr.1:113894194-113905202 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP4B1 | NM_001253852.2 | 2 | 368 | NP_001240781.1 |
| NM_001253853.2 | NP_001240782.1 | |||
| NM_001308312.1 | NP_001295241.1 | |||
| NM_006594.4 | 3 | 421 | NP_006585.2 | |
| XM_011540523.2 | XP_011538825.1 | |||
| XM_011540524.2 | XP_011538826.1 | |||
| XM_011540525.2 | 3 | 499 | XP_011538827.1 | |
| XM_011540528.2 | 2 | 2105 | XP_011538830.1 | |
| XM_017000088.1 | 2 | 2105 | XP_016855577.1 | |
| XM_017000089.1 | 2 | 216 | XP_016855578.1 | |
| XM_017000090.1 | XP_016855579.1 | |||
| XM_017000091.1 | 3 | 499 | XP_016855580.1 | |
| XM_017000092.1 | 2 | 216 | XP_016855581.1 | |
| XM_017000093.1 | 2 | 216 | XP_016855582.1 | |
| AB209817.1 | 2 | 305 | BAD93054.1 | |
| AF092094.1 | 3 | 365 | AAD20448.1 | |
| AK093740.1 | ||||
| AK298037.1 | 2 | 308 | ||
| AK300204.1 | ||||
| AK314641.1 | 3 | 325 | ||
| BC014146.2 | 2 | 226 | AAH14146.1 | |
| BG718098.1 | ||||
| DC299853.1 | 2 | 348 | ||
| DC367373.1 | 3 | 417 | ||
| DC368382.1 | 2 | 368 | ||
| DC390197.1 | 3 | 403 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587251 | Chr.1:113871703 - 113936287 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| nsv819786 | Chr.1:113901490 - 113903616 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map