Assay Details
Target Gene Details
Entrez Gene ID: | 606495 |
Gene Name: | cytochrome b5 reductase like |
Gene Aliases: |
- |
Location: |
Chr.1:54172336-54200073 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5RL | NM_001031672.2 | 3 | 419 | NP_001026842.2 |
| AK090456.1 | 3 | 357 | BAC03437.1 | |
| CD515193.1 | 1 | 165 | ||
| DA793739.1 | 3 | 419 |
More Information
Additional Information:
For this assay, SNP(s) [rs79743819] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map