Assay Details
Target Gene Details
Entrez Gene ID: | 10542 |
Gene Name: | late endosomal/lysosomal adaptor, MAPK and MTOR activator 5 |
Gene Aliases: |
HBXIP, XIP |
Location: |
Chr.1:110401253-110407924 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMTOR5 | NM_006402.2 | NP_006393.2 | ||
| AA443738.1 | ||||
| AF029890.1 | AAC52032.1 | |||
| AK054769.1 | ||||
| AK056679.1 | ||||
| BC062619.1 | AAH62619.2 |
More Information
Set Membership: |
|

Genomic Map