Assay Details
Target Gene Details
Entrez Gene ID: | 7156 |
Gene Name: | topoisomerase (DNA) III alpha |
Gene Aliases: |
TOP3, ZGRF7 |
Location: |
Chr.17:18269958-18315092 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOP3A | NM_001320759.1 | 18 | 3721 | NP_001307688.1 |
| NM_004618.4 | 19 | 3795 | NP_004609.1 | |
| XM_011524001.2 | 14 | 2886 | XP_011522303.1 | |
| AF086042.1 | 1 | 138 | ||
| AK126869.1 | 13 | 2836 | ||
| AK290513.1 | 8 | 2106 | ||
| AK292926.1 | ||||
| BC051748.1 | 19 | 3677 | AAH51748.1 | |
| BY996313.1 | 1 | 472 | ||
| U43431.1 | 19 | 3658 | AAB03694.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833386 | Chr.17:18266947 - 18429412 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map