Assay Details
Target Gene Details
Entrez Gene ID: | 79885 |
Gene Name: | histone deacetylase 11 |
Gene Aliases: |
HD11 |
Location: |
Chr.3:13480171-13506424 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDAC11 | NM_001136041.2 | NP_001129513.1 | ||
| NM_024827.3 | NP_079103.2 | |||
| XM_005265491.1 | XP_005265548.1 | |||
| XM_005265492.1 | XP_005265549.1 | |||
| XM_006713338.1 | XP_006713401.1 | |||
| XM_011534131.1 | XP_011532433.1 | |||
| XM_011534132.2 | XP_011532434.2 | |||
| XM_011534135.1 | XP_011532437.1 | |||
| XM_017007234.1 | XP_016862723.1 | |||
| AK025426.1 | BAB15127.1 | |||
| AK025890.1 | BAB15272.1 | |||
| AK293223.1 | ||||
| AK299021.1 | ||||
| AL834223.1 | CAH56375.1 | |||
| BC009676.1 | AAH09676.1 | |||
| BG708402.1 | ||||
| DB025431.1 |
More Information
Set Membership: |
|

Genomic Map