Assay Details
Target Gene Details
Entrez Gene ID: | 23345 |
Gene Name: | spectrin repeat containing nuclear envelope protein 1 |
Gene Aliases: |
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2 |
Location: |
Chr.6:152121684-152637399 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 60 - Exon 60 |
Target Gene Details
Entrez Gene ID: | 100505475 |
Gene Name: | SYNE1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:152380530-152381713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNE1-AS1 | NR_120501.1 | |||
| CK821189.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015648 | Chr.6:152339175 - 152508657 on Build GRCh38 | Loss |
|
| nsv1032029 | Chr.6:152166450 - 152408521 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map